missing translation for 'onlineSavingsMsg'
Learn More

Aminoacetonitrile bisulfate, 98%, Thermo Scientific Chemicals

Catalog Number 10542361
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
151-63-3
C2H4N2·H2SO4
154.14
MFCD00012851
GTGIXCPOLMWQTC-UHFFFAOYSA-N
aminoacetonitrile bisulfate, 2-aminoacetonitrile sulfate, aminoacetonitrile hydrogen sulfate, aminoacetonitrile sulfate 1:1, aminoacetonitrile hydrogensulfate, 2-aminoacetonitrile; sulfuric acid, aminoacetonitrile bisulphate, unii-7xa82hgq1c, aminoacetonitrile hydrosulfate, aminomethylcyanide hydrogen sulfate
196971
2-aminoacetonitrile;sulfuric acid
C(C#N)N.OS(=O)(=O)O
This item is not returnable. View return policy
151-63-3
C2H4N2·H2SO4
154.14
MFCD00012851
GTGIXCPOLMWQTC-UHFFFAOYSA-N
aminoacetonitrile bisulfate, 2-aminoacetonitrile sulfate, aminoacetonitrile hydrogen sulfate, aminoacetonitrile sulfate 1:1, aminoacetonitrile hydrogensulfate, 2-aminoacetonitrile; sulfuric acid, aminoacetonitrile bisulphate, unii-7xa82hgq1c, aminoacetonitrile hydrosulfate, aminomethylcyanide hydrogen sulfate
196971
2-aminoacetonitrile;sulfuric acid
C(C#N)N.OS(=O)(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 123.0°C to 125.0°C
Color Beige-Yellow to White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula H2NCH2CN·H2SO4
Quantity 100 g
Beilstein 04,344
Merck Index 15,406
Formula Weight 154.14
Percent Purity 98%
Physical Form Crystalline Powder and/or Chunks
Chemical Name or Material Aminoacetonitrile bisulfate
Show More Show Less
compliance-icons
Product Identifier
  • Aminoacetonitrile bisulfate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'