Learn More
Amikacin M.I.C.Evaluator Strips (M.I.C.E.™)
Determine accurate Minimum Inhibitory Concentration (MIC) values of antibiotics using MIC testing strips.
Brand:  Thermo Scientific Oxoid MA0135D
Code : 50
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 13245279
  • £41.97 / Pack of 10
Estimated Shipment
04-06-2018
Add to basket
Description
Description
Conveniently packaged M.I.C.E. strips combine the ease of a diffusion test with the accuracy of an MIC test.
Simple to Perform
  • Distinctive gradient format provides excellent contrast with the agar
  • Increased font size makes reading easier
  • Scale conforms to international standards
  • Strips are tailored to meet clinical requirements while maintaining the accuracy required for research
Easy-to-handle
  • The strip is conveniently presented when the sachet is peeled apart
  • Single-strip packaging eliminates the requirement to maintain and quality control opened strips; open one and use it
  • Stability of the individual M.I.C.E. strips is ensured through the inclusion of a desiccant within each sachet
  • 10 and 50 pack sizes offer users maximum flexibility to put together custom panels for testing
  • Easily stackable boxes simplify storage; all antimicrobials are stored in the fridge (2-8°C)
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
Specifications
AK
10 Strips/Pk.
256-0.015µg/mL
Amikacin M.I.C.Evaluator Strips (M.I.C.E™)
2-8°C
Videos