missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse AMIGO3 Antibody, R&D Systems™
Goat Polyclonal Antibody
POA
Specifications
Antigen | AMIGO3 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready |
Applications | Western Blot, Flow Cytometry, CyTOF |
Classification | Polyclonal |
Description
AMIGO3 Polyclonal specifically detects AMIGO3 in Mouse samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.Specifications
AMIGO3 | |
Western Blot 0.1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
Polyclonal | |
Goat | |
Mouse | |
adhesion molecule with Ig-like domain 3, ALI3, Alivin-3, AMIGO-3, amphoterin-induced gene and ORF 3, amphoterin-induced protein 3, KIAA1851, MGC120552 | |
AMIGO3 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot, Flow Cytometry, CyTOF | |
Unconjugated | |
RUO | |
Q8C2S7 | |
386724 | |
Mouse myeloma cell line NS0-derived recombinant mouse AMIGO3 Thr20-Pro378 Accession # Q8C2S7 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects mouse AMIGO3 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 5% cross-reactivity with recombinant mouse AMIGO2 is observed and less than 2% cross-reactivity with recombinant human AMIGO1 is observed. |
For Research Use Only