missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human AMIGO Antibody, R&D Systems™
Sheep Polyclonal Antibody
POA
Specifications
Antigen | AMIGO |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Description
AMIGO Polyclonal specifically detects AMIGO in Human samples. It is validated for Western Blot.Specifications
AMIGO | |
Western Blot 0.1 ug/mL | |
Polyclonal | |
Sheep | |
Human | |
adhesion molecule with Ig-like domain 1, ALI2, Alivin-2, AMIGO1, AMIGO-1, AMIGOamphoterin-induced protein 1, amphoterin-induced gene and ORF, KIAA1163 | |
AMIGO1 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
Q86WK6 | |
57463 | |
Mouse myeloma cell line NS0-derived recombinant human AMIGO Met1-Gly365 Accession # Q86WK6 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human AMIGO in direct ELISAs and Western blots. In direct ELISAs, approximately 5% cross-reactivity with recombinant human (rh) AMIGO2 is observed and less than 1% cross-reactivity with rhAMIGO3 is observed. |
For Research Use Only