Learn More
Amidosulfonic acid, 98+%, Thermo Scientific Chemicals
Description
Amidosulfonic acid is widely utilized as a green catalyst for the preparation of amide from ketoxime. It finds an application as a titrant in the determination of the burette injection volume and chemical calibration factor. It is an efficient catalyst in the synthesis of polyhydroquinoline derivatives by Hantzsch condensation reaction, deazaoxaflavin, fatty acid methyl and ethyl esters and in the isolation of 5-hydroxymethylfurfural (HMF) from bamboo fiber. It is involved in the determination of silicates in water samples. It acts as a neutralizing agent during the determination of paracetamol by Glynn and Kendal colorimetric method.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Amidosulfonic acid is widely utilized as a green catalyst for the preparation of amide from ketoxime. It finds an application as a titrant in the determination of the burette injection volume and chemical calibration factor. It is an efficient catalyst in the synthesis of polyhydroquinoline derivatives by Hantzsch condensation reaction, deazaoxaflavin, fatty acid methyl and ethyl esters and in the isolation of 5-hydroxymethylfurfural (HMF) from bamboo fiber. It is involved in the determination of silicates in water samples. It acts as a neutralizing agent during the determination of paracetamol by Glynn and Kendal colorimetric method.
Solubility
Slightly soluble in water, dimethyl fluoride and methanol. Insoluble in hydrocarbons.
Notes
Incompatible with chlorine compounds, nitrates, carbonates, strong oxidizing agents and strong bases.
Specifications
pH | 1.2 |
Melting Point | ∼205°C (decomposition) |
Density | 2.12 |
Odor | Odorless |
Quantity | 100 g |
Linear Formula | H2NSO3H |
UN Number | UN2967 |
Merck Index | 148921 |
Solubility Information | Slightly soluble in water,dimethyl fluoride and methanol. Insoluble in hydrocarbons. |
Formula Weight | 97.09 |
Show More |
RUO – Research Use Only