missing translation for 'onlineSavingsMsg'
Learn More

Amidinothiourea, 99+%, Thermo Scientific Chemicals

Catalog Number p-56097
Click to view available options
Quantity:
10 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
2114-02-5
C2H6N4S
118.16
MFCD00014472
OKGXJRGLYVRVNE-UHFFFAOYSA-N
amidinothiourea, guanylthiourea, guthimin, guthimine, 2-imino-4-thiobiuret, n-amidinothiourea, thiourea, aminoiminomethyl, thiodicyanodiamidine, diaminomethylidene thiourea, gutimine
2724563
diaminomethylidenethiourea
C(=NC(=S)N)(N)N
This item is not returnable. View return policy
2114-02-5
C2H6N4S
118.16
MFCD00014472
OKGXJRGLYVRVNE-UHFFFAOYSA-N
amidinothiourea, guanylthiourea, guthimin, guthimine, 2-imino-4-thiobiuret, n-amidinothiourea, thiourea, aminoiminomethyl, thiodicyanodiamidine, diaminomethylidene thiourea, gutimine
2724563
diaminomethylidenethiourea
C(=NC(=S)N)(N)N

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 160°C to 164°C
Color White
Loss on Drying 0.3% max.
Infrared Spectrum Authentic
Assay Percent Range 99% min. (Iodimetry)
Packaging Glass bottle
Linear Formula NH2C(=NH)NHC(=S)NH2
Quantity 10 g
Solubility Information Solubility in water: 7 w/w % in water (20°C)
Formula Weight 118.16
Percent Purity 99+%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Amidinothiourea, 99+%
Show More Show Less
compliance-icons
Product Identifier
  • Amidinothiourea
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'