Learn More
EVOS™ FL Auto Cell Imaging System Accessory, Onstage Incubator Vessel Holders
Brand:  EVOS™ AMEPVH030
Code : NEW
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 15250211
  • £212.49 / Each
Estimated Shipment
18-06-2018
Add to basket
Description
Description
For use with EVOS FL Auto Digital Inverted Fluorescence System (Cat. No. 12-463-377).
Onstage Vessel Holders
  • Accommodate a variety of flasks, plates and dishes
  • Maximize your choice of cell culture vessels when conducting time-lapse imaging experiments using the EVOS FL Auto Imaging System equipped with onstage incubator
  • Allow a perfect fit of your microwell plates or cell culture flasks/dishes to the master plate of the onstage incubator for increased precision in sample alignment
  • With a snug and secure fit, yet readily switched, these vessel holders make time-lapse imaging convenient and easy
Specifications
Specifications
non-cGMP
EVOS™ Onstage incubator, two 35mm petri dish
Two 35mm Petri dishes
Videos