missing translation for 'onlineSavingsMsg'
Learn More

AMG 9810, Tocris Bioscience™

Potent and selective, competitive antagonist of TRPV1

Brand:  Tocris (Bio-Techne) 2316/10

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13461014

  • £166.11 / 10mg

Estimated Shipment date
from Supplier 25-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Potent and selective, competitive vanilloid TRPV1 receptor antagonist (IC50 = 17 nM). Inhibits capsaicin-, proton-, heat- and endogenous ligand-induced activation of human and rat recombinant TRPV1 receptors. Displays antihyperalgesic properties in a rat model of inflammatory pain.
Specifications

Chemical Identifiers

545395-94-6
337.419
unii-182hij2d7f, 2e-3-4-tert-butylphenyl-n-2,3-dihydro-1,4-benzodioxin-6-yl prop-2-enamide, 2e-n-2,3-dihydro-1,4-benzodioxin-6-yl-3-4-1,1-dimethylethyl phenyl-2-propenamide, trans-cinnamide, d0xo2g, e-3-4-t-butylphenyl-n-2,3-dihydrobenzo b 1,4 dioxin-6-yl acrylamide, 3-4-t-butylphenyl-n-2,3-dihydrobenzo b 1,4 dioxin-6-yl acrylamide, amg hplc , powder, e-3-4-tert-butylphenyl-n-2,3-dihydro-1,4-benzodioxin-6-yl prop-2-enamide
(E)-3-(4-tert-butylphenyl)-N-(2,3-dihydro-1,4-benzodioxin-6-yl)prop-2-enamide
C21H23NO3
GZTFUVZVLYUPRG-IZZDOVSWSA-N
680502
CC(C)(C)C1=CC=C(C=C1)C=CC(=O)NC2=CC3=C(C=C2)OCCO3

Specifications

10mg
545395-94-6
unii-182hij2d7f, 2e-3-4-tert-butylphenyl-n-2,3-dihydro-1,4-benzodioxin-6-yl prop-2-enamide, 2e-n-2,3-dihydro-1,4-benzodioxin-6-yl-3-4-1,1-dimethylethyl phenyl-2-propenamide, trans-cinnamide, d0xo2g, e-3-4-t-butylphenyl-n-2,3-dihydrobenzo b 1,4 dioxin-6-yl acrylamide, 3-4-t-butylphenyl-n-2,3-dihydrobenzo b 1,4 dioxin-6-yl acrylamide, amg hplc , powder, e-3-4-tert-butylphenyl-n-2,3-dihydro-1,4-benzodioxin-6-yl prop-2-enamide
CC(C)(C)C1=CC=C(C=C1)C=CC(=O)NC2=CC3=C(C=C2)OCCO3
337.419
337.42
AMG 9810
C21H23NO3
GZTFUVZVLYUPRG-IZZDOVSWSA-N
(E)-3-(4-tert-butylphenyl)-N-(2,3-dihydro-1,4-benzodioxin-6-yl)prop-2-enamide
680502
>98%
Videos
Documents