missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Amersham™ Megaprime™ DNA Labeling System
For labeling DNA probes to specific activities >1 x 109dpm/μg in 10 minutes
Brand: Cytiva RPN1606
Code : GE
Additional Details : Weight : 0.12200kg
Includes: Primer solution; Labeling buffer; Reaction buffer; Enzyme; Standard DNA solution; Carrier DNA solution
Description
Megaprime™ DNA Labeling Systems are used for rapid, random-prime labeling from as little as 25ng substrate DNA.
- Features cloned Klenow fragment and random nonamers in the random-prime labeling technique
- No need to remove unincorporated nucleotides before hybridization
- Protocols for labeling directly from low melting point agarose
Specifications
-15°C to -30°C | |
Primer solution 150μL, Labeling Buffer dATP, dGTP and dTTP in Tris HCl pH7.5, 2-mercaptoethanol and MgCl2 300μL, Nucleotide solutions dATP, cCTP, dGTP, dTTP in Tris HCl pH 8, 0.5mM EDTA, Reaction Buffer A 10 x concentra | |
Megaprime DNA Labeling System, dCTP (for use with labeled dCTP) |
Membrane hybridization | |
30 |