missing translation for 'onlineSavingsMsg'
Learn More

AMD 3465 hexahydrobromide, Tocris Bioscience™

Potent and selective CXCR4 antagonist

Brand:  RD Systems (Bio-Techne) 4179/50

Code : Z1

Additional Details : Weight : 0.00050kg

Quantity 50mg

 View more versions of this product

Product Code. 13444294

  • £818.79 / 50mg

Estimated Shipment date
from Supplier 31-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Potent, selective CXCR4 antagonist; exhibits 8-fold higher affinity than AMD 3100 (Cat.No. 3299). Inhibits SDF-1α-ligand binding (Ki = 41.7 nM). Potently inhibits HIV cell entry in vitro; causes leukocytosis and mobilizes haematopoietic stem cells in vivo.
Specifications

Chemical Identifiers

185991-07-5
896.082
amd 3465 hexahydrobromide, pyridin-2-ylmethyl 4-1,4,8,11-tetraazacyclotetradecan-1-ylmethyl phenyl methyl amine hexahydrobromide, n-4-1,4,8,11-tetraazacyclotetradec-1-ylmethyl phenyl methyl-2-pyridinemethanamine hexahydrobromide, amd3465 hexahydrobromide, amd3465 6 hbr, n-pyridin-2-ylmethyl-1-4-1,4,8,11-tetrazacyclotetradec-1-ylmethyl phenyl methanamine hexahydrobromide
N-(pyridin-2-ylmethyl)-1-[4-(1,4,8,11-tetrazacyclotetradec-1-ylmethyl)phenyl]methanamine;hexahydrobromide
C24H44Br6N6
ARHBIBDGWDRBJH-UHFFFAOYSA-N
9897616
C1CNCCNCCCN(CCNC1)CC2=CC=C(C=C2)CNCC3=CC=CC=N3.Br.Br.Br.Br.Br.Br

Specifications

AMD 3465 hexahydrobromide
185991-07-5
amd 3465 hexahydrobromide, pyridin-2-ylmethyl 4-1,4,8,11-tetraazacyclotetradecan-1-ylmethyl phenyl methyl amine hexahydrobromide, n-4-1,4,8,11-tetraazacyclotetradec-1-ylmethyl phenyl methyl-2-pyridinemethanamine hexahydrobromide, amd3465 hexahydrobromide, amd3465 6 hbr, n-pyridin-2-ylmethyl-1-4-1,4,8,11-tetrazacyclotetradec-1-ylmethyl phenyl methanamine hexahydrobromide
C1CNCCNCCCN(CCNC1)CC2=CC=C(C=C2)CNCC3=CC=CC=N3.Br.Br.Br.Br.Br.Br
896.082
896.07
50mg
C24H44Br6N6
ARHBIBDGWDRBJH-UHFFFAOYSA-N
N-(pyridin-2-ylmethyl)-1-[4-(1,4,8,11-tetrazacyclotetradec-1-ylmethyl)phenyl]methanamine;hexahydrobromide
9897616
>97%
Videos
Documents