Learn More
Invitrogen™ RPA III™ Ribonuclease Protection Assay Kit
Fast single-tube reaction
Brand:  Invitrogen™ AM1415
Code : 50
Additional Details : Weight : 0.09000kg
 View more versions of this product
Product Code. 10117304
  • £186.45 / Each
Estimated Shipment
09-02-2018
Add to basket
For Research Use Only. All usage must comply with product instructions.
Disclaimers:
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Specifications
Kit without manual
35 reactions
Store Hybridization III Buffer, RNase Digestion III Buffer, Yeast RNA, RNase Inactivation/Precipitation III Solution, Gel Loading Buffer II, pTRI-Actin-Mouse, RNase A/RNase T1 Mix, RNase T1, and 5 M Ammonium Acetate at -20°C. Store Mouse Liver RNA at below -70°C. Store Probe Elution Buffer at 4°C.
Videos