Learn More
Invitrogen™ RPA III™ Ribonuclease Protection Assay Kit
Fast single-tube reaction
Brand:  Invitrogen™ AM1415M
Code : 50
Additional Details : Weight : 0.20000kg
 View more versions of this product
Product Code. 10269534
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
Kit with manual
35 reactions
Store Hybridization III Buffer, RNase Digestion III Buffer, Yeast RNA, RNase Inactivation/Precipitation III Solution, Gel Loading Buffer II, pTRI-Actin-Mouse, RNase A/RNase T1 Mix, RNase T1, and 5 M Ammonium Acetate at -20°C. Store Mouse Liver RNA at below -70°C. Store Probe Elution Buffer at 4°C.
For Research Use Only. Not for use in diagnostic procedures.