Learn More
Invitrogen™ RNAqueous-Midi Total RNA Isolation Kit
RNA isolation for midrange amounts of starting material
Brand:  Invitrogen™ AM1911M
Code : 50
Additional Details : Weight : 1.13000kg
 View more versions of this product
Product Code. 10187674
  • £217.36 / Each
Estimated Shipment
05-02-2018
Add to basket
For Research Use Only. All usage must comply with product instructions.
Disclaimers:
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Specifications
Total RNA
Formaldehyde Load Dye, Lysis/Binding Solution, Water for 64% Ethanol, Lithium, Chloride Solution, Wash Solution No. 1 and Wash Solution 2/3 Concentrate, Syringe Filter Units, Collection Tubes and Elution Solution, Manual
Kit with manual
Tissue Cells
Northern Blotting, Nuclease Protection Assays, RT-PCR (Endpoint), Real-Time PCR, cDNA Library Construction
Syringe Filter (Glass Fiber)
15 preps
Total RNA
Videos