missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RNA Rapid Extraction Solution

Fast procedure to release RNA from Whatman FTA cards

£97.70 - £100.00

Specifications

For Use With (Application) Northern Blotting, RT-PCR (Endpoint), Real-Time PCR
Quantity 10mL
Storage Requirements Store at room temperature.
View More Specs

Products 2
Product Code Brand Includes Price Quantity & Availability  
Product Code Brand Includes Price Quantity & Availability  
10056004 View Documents Invitrogen™
AM9775M
Solution, with manual £97.70
Each View alternative products
Save Up to 
»
Estimated Shipment
23-07-2020
Please to purchase this item. Need a web account? Register with us today!
10311525 View Documents Invitrogen™
AM9775
Solution, without manual £100.00
Each View alternative products
Save Up to 
»
In stock, ready to ship: 12 Please to purchase this item. Need a web account? Register with us today!
Description

Description

Invitrogen™ RNA Rapid Extraction Solution is to release RNA from the paper prior to RNA isolation. 10 mL is provided.
  • Rapid 5 min procedure to release RNA from Wha™an™ FTA™ cards
  • Ideal for high-throughput applications
  • Integrity of the RNA is unaffected
Ready for real-time PCR Disks punched from FTA™ Cards are simply dropped into RNA Rapid Extraction Solution and agitated for 5 minutes to release the sample for RNA isolation—while protecting the RNA from degradation. The RNA can then be purified from the RNA Rapid Extraction Solution using the MagMAX™-96 Viral RNA Isolation Kit (SKU #AM1836). The high-quality purified RNA is suitable for quantitative RT-PCR or other downstream RNA applications. Wha™an™ FTA™ Cards can be used with virtually any sample type including blood, cultured cells, buccal cells, plant material, bacteria, plasmids, microorganisms, solid tissue, viral particles, and more.

Order Info

Shipping Condition: Room temperature

Specifications

Specifications

Northern Blotting, RT-PCR (Endpoint), Real-Time PCR
10mL
Store at room temperature.

For Research Use Only. Not for use in diagnostic procedures.