Learn More
Invitrogen™ RNA Rapid Extraction Solution
Fast procedure to release RNA from Whatman FTA cards
Brand:  Invitrogen™ AM9775M
Code : 50
Additional Details : Weight : 0.10500kg
 View more versions of this product
Product Code. 10056004
  • £97.70 / Each
Estimated Shipment date
from Supplier 12-06-2020
For Research Use Only. All usage must comply with product instructions.
Description
Description
Invitrogen™ RNA Rapid Extraction Solution is to release RNA from the paper prior to RNA isolation. 10 mL is provided.
  • Rapid 5 min procedure to release RNA from Wha™an™ FTA™ cards
  • Ideal for high-throughput applications
  • Integrity of the RNA is unaffected
Ready for real-time PCR Disks punched from FTA™ Cards are simply dropped into RNA Rapid Extraction Solution and agitated for 5 minutes to release the sample for RNA isolation—while protecting the RNA from degradation. The RNA can then be purified from the RNA Rapid Extraction Solution using the MagMAX™-96 Viral RNA Isolation Kit (SKU #AM1836). The high-quality purified RNA is suitable for quantitative RT-PCR or other downstream RNA applications. Wha™an™ FTA™ Cards can be used with virtually any sample type including blood, cultured cells, buccal cells, plant material, bacteria, plasmids, microorganisms, solid tissue, viral particles, and more.
Specifications
Specifications
Northern Blotting, RT-PCR (Endpoint), Real-Time PCR
Solution, with manual
10mL
Store at room temperature.