Learn More
Invitrogen™ QuantumRNA™ β-Actin Internal Standards
Used for quantitative end-point RT-PCR
Brand:  Invitrogen™ AM1720M
Code : 50
Additional Details : Weight : 0.03000kg
 View more versions of this product
Product Code. 10361225
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
Specifications
Specifications
Kit with manual
100 reactions
Store β-Actin Primer Pair and β-Actin Competimers at -20°C. Store Control Template RNA below -70°C. Store Nuclease-free water at any temperature.