Invitrogen™ Ambion™ mMESSAGE mMACHINE™ T7 ULTRA Transcription Kit

Generate RNA transcripts with higher protein yields

Overview
Brand: Invitrogen™

Manufacturer Part Number: AM1345M

UNSPSC: 41105804

Code: 50

Additional Details:
Additional Details: Weight: 0.07000kg



Disclaimers: For Research Use Only. Not for use in diagnostic procedures.

Product Code. 10418834

Quantity Price
1 £ 391.0 / Each
Estimated Shipment date
from Supplier 11-05-2017
Add to basket

For Research Use Only. All usage must comply with product instructions.
 

Description and Specification

Specification

Includes T7 Enzyme Mix, 10X Reaction Buffer, 2XNTP/ARCA Solution, GTP Solution, pTRI-Xef, TURBO DNase, Ammonium Acetate Stop Solution, Lithium Chloride Precipitation Solution, E-PAP, 5X E-PAP Buffer, ATP Solution, MnCl2, Formaldehyde Load Dye, and Nuclease-free Water
Concentration 40mM
Product Type Kit with manual
For Use With (Application) Gene Expression Analysis and Genotyping, in vitro Transcription
Quantity 10 reactions