Invitrogen™ Ambion™ mMESSAGE mMACHINE™ T3 Transcription Kit
Obtain 10–50 times yield of conventional in vitro transcription reactions
Overview
Brand: Invitrogen™
Manufacturer Part Number: AM1348M
UNSPSC: 41105804
Code: 50
Additional Details:
Additional Details: Weight: 0.07000kg
Disclaimers: For Research Use Only. Not for use in diagnostic procedures.
Product Code. 10187294
Quantity Price
1 £ 390.0 / Each
Estimated Shipment date
from Supplier 12-04-2017
Add to basket
For Research Use Only. All usage must comply with product instructions.
Description and Specification
Specification
Product Type Kit with manual
Includes T3 Enzyme Mix, 10X Reaction Buffer, 2XNTP/CAP Solution, GTP Solution, pTRI-Xef, TURBO DNase, Ammonium Acetate Stop Solution, Lithium Chloride Precipitation Solution, Gel Loading Buffer II, and Nuclease-free Water
For Use With (Application) Gene Expression Analysis and Genotyping, in vitro Transcription
Quantity 25 reactions