Learn More
Invitrogen™ Microcentrifuge Spin Columns
The microcentrifuge spin columns included with the FastTrack™ 2.0 and Micro-FastTrack 2.0 Kits are available separately.
Brand: Invitrogen™ R50020
972.04 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Code : 50
Additional Details : Weight : 0.20000kg
Description
The microcentrifuge spin columns included with the FastTrack™ 2.0 and Micro-FastTrack 2.0 Kits are available separately. The columns are made from polypropylene and are designed to enhance the speed and efficiency of polyA+ RNA isolation. The columns are sterilized and RNase-free.
Specifications
0.45 micron cellulose acetate microcentrifuge spin columns and sterile microcentrifuge tubes. After opening, store microcentrifuge columns and tubes in a sterile container. | |
200 Columns |
For Research Use Only. Not for use in diagnostic procedures.