Learn More
Invitrogen™ MEGAscript™ T3 Transcription Kit
Synthesize 10 to 50 times the amount of RNA produced by conventional transcription reactions
Brand:  Invitrogen™ AM1338M
Code : 50
Additional Details : Weight : 0.07000kg
 View more versions of this product
Product Code. 10782687
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
Specifications
Specifications
Gene Expression Analysis and Genotyping, in vitro Transcription
T3 Enzyme Mix, 10X Reaction Buffer, ATP Solution, CTP Solution, GTP Solution, UTP Solution, pTRI-Xef, TURBO DNase, Ammonium Acetate Stop Solution, Lithium Chloride Precipitation Solution, Gel Loading Buffer II, and Nuclease-free Water
Kit with manual
40 reactions