Invitrogen™ Ambion™ MAXIscript™ SP6/T7 Transcription Kit
Fast synthesis of RNA probes for use in ribonuclease protection assays, in situ hybridization, and blot hybridizations
Overview
Brand: Invitrogen™
Manufacturer Part Number: AM1322M
100 RXN MAXISCRIPT SP6/T7 KIT (50 RXNS EACH) WITH
UNSPSC: 41105804
Code: 50
Additional Details:
Additional Details: Weight: 0.00450kg
Disclaimers: For Research Use Only. Not for use in diagnostic procedures.
Product Code. 10794027
Quantity Price
1 £ 286.0 / Each
Estimated Shipment date
from Supplier 10-03-2017
Add to basket
For Research Use Only. All usage must comply with product instructions.
Description and Specification
Specification
Product Name Kit with manual
Product Type Kit with manual
Quantity 100 reactions
For Use With (Application) DNA and RNA Purification and Analysis, Gene Expression Analysis and Genotyping, in vitro Transcription, Northern Blotting, Nuclease Protection Assays, Nucleic Acid Gel Electrophoresis and Blotting, Southern Blotting
Includes SP6 Enzyme Mix, T7 Enzyme Mix, 10X Transcription Buffer, ATP Solution, CTP Solution, GTP Solution, UTP Solution, TURBO DNase, Gel Loading Buffer II, pTRI-b-actin, and Nuclease-free Water
Quantity 100 reactions