missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ M-MLV Reverse Transcriptase
Commonly used in the first step of RT-PCR as well as for primer extension experiments
£93.28 - £206.23
Specifications
For Use With (Application) | RT-PCR |
---|---|
Sample Type | ssRNA, ssDNA, RNA-DNA Hybrid |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10596925
|
Invitrogen™
AM2043 |
4000 units |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
10350545
|
Invitrogen™
AM2044 |
10,000 units |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Moloney Murine Leukemia Virus Reverse Transcriptase (M-MLV RT) is derived from a cloned region of the pol gene of M-MLV and isolated from an E. coli strain overexpressing this construct.
- In the presence of a single-stranded RNA or DNA template, primer, and deoxyribonucleotides, M-MLV Reverse Transcriptase (RT) will extend the oligonucleotide primer, producing a complementary strand
- Optimal reaction temperature: 37°C
- Final product size: 7kb or less
PCR & Real-Time PCR, Reverse Transcription
Order Info
Shipping Condition: Dry ice
Specifications
RT-PCR | |
ssRNA, ssDNA, RNA-DNA Hybrid |
For Research Use Only. Not for use in diagnostic procedures.