Learn More
Invitrogen™ Guanidine Thiocyanate
Guanidine Thiocyanate is an ultrapure, molecular biology grade reagent. It is free of detectable nuclease and protease activity and is a strong protein denaturant, as both the guanidinium cation and the thiocyanate anion are chaotropic agents.
Brand: Invitrogen™ AM9422
Code : 50
Additional Details : Weight : 1.20000kg Transport : UN number : 1759 Chem class : 8 Pack group : III
Description
Guanidine Thiocyanate is an ultrapure, molecular biology grade reagent. It is free of detectable nuclease and protease activity and is a strong protein denaturant, as both the guanidinium cation and the thiocyanate anion are chaotropic agents. It is provided in one bottle containing 500 g. It is recommended for isolation of RNA, especially for tissues such as pancreas with high levels of RNase activity. In solution, it is known as guanidinium thiocyanate. Guanidine Thiocyanate is thoroughly tested for contaminating nonspecific endonuclease, exonuclease, and RNase activity.
Order Info
Shipping Condition: Room temperature
Specifications
Store at room temperature. | |
Guanidine Isothiocyanate | |
>99% | |
Room Temperature |
RNA Extraction | |
Ambion™ | |
500 g | |
Powder |
For Research Use Only. Not for use in diagnostic procedures.