Learn More
Invitrogen™ Barrier (Filter) Tips, 1,000 μL size
Aerosol filter prevents cross-contamination of samples
Brand: Invitrogen™ AM12665
Code : 50
Additional Details : CAS Number : 9048-46-8 Weight : 2.58000kg
Includes: Ten racks containing 100 tips
Description
Compatible with Eppendorf™, Finnpipette, Oxford Benchmate™, Pipetman™, Proline™, Rainin, Socorex, and other brands of pipetters
- Invitrogen Barrier tips prevent cross-contamination of samples by means of aerosol filter
- They are RNase- and DNase-free and are ideal for handling RNA, PCR applications, and for working with radioisotopes or toxic chemicals
- Each lot of tips is subjected to rigorous testing for RNase contamination and is certified to be nuclease-free
DNA & RNA Purification & Analysis, General RNA Purification Reagents & Accessories, PCR, PCR & Real-Time PCR, RNA Extraction, Real Time PCR (qPCR), Total RNA Isolation, Total RNA from Animal Cells & Tissues, Transcriptome RNA Isolation, mRNA Isolation, microRNA Isolation
Specifications
1000 μL | |
Sterile | |
Barrier | |
Room Temperature | |
Invitrogen |
Filtered | |
Rack | |
Store at room temperature. | |
Universal Fit |
For Research Use Only. Not for use in diagnostic procedures.