missing translation for 'onlineSavingsMsg'
Learn More

Amberlyst™ 15(wet) ion-exchange resin, ACROS Organics™

£11.33 - £169.46

Chemical Identifiers

CAS 9037-24-5
Molecular Formula C3H7BrO
Molecular Weight (g/mol) 138.992
MDL Number MFCD00145841
InChI Key YZUPZGFPHUVJKC-UHFFFAOYSA-N
Synonym 2-bromoethyl methyl ether, 2-methoxyethyl bromide, ethane, 1-bromo-2-methoxy, ether, 2-bromoethyl methyl, methoxyethyl bromide, 2-bromoethylmethylether, bromoethyl methyl ether, 2-bromoethylmethyl ether, 2-bromoethylmethyl-ether, 2-bromo-1-methoxyethane
PubChem CID 80972
IUPAC Name 1-bromo-2-methoxyethane
SMILES COCCBr
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10543951
SDS View Documents
Acros Organics
202150250
25g Plastic bottle £11.33
25g View alternative products
Save Up to 
»
In stock, ready to ship: 16 Please to purchase this item. Need a web account? Register with us today!
10114270
SDS View Documents
Acros Organics
202155000
500g Plastic bottle £21.51
500g View alternative products
Save Up to 
»
In stock, ready to ship: 7 Please to purchase this item. Need a web account? Register with us today!
13448448
View Documents
Acros Organics
202150050
5kg Plastic drum £169.46
5kg View alternative products
Save Up to 
»
Estimated Shipment
09-11-2020
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Ion exchange resin
Specifications

Chemical Identifiers

9037-24-5
138.992
YZUPZGFPHUVJKC-UHFFFAOYSA-N
80972
COCCBr
C3H7BrO
MFCD00145841
2-bromoethyl methyl ether, 2-methoxyethyl bromide, ethane, 1-bromo-2-methoxy, ether, 2-bromoethyl methyl, methoxyethyl bromide, 2-bromoethylmethylether, bromoethyl methyl ether, 2-bromoethylmethyl ether, 2-bromoethylmethyl-ether, 2-bromo-1-methoxyethane
1-bromo-2-methoxyethane

Specifications

Amberlyst™ 15
9037-24-5
Particle size analysis: uniformity coefficient: <1.70, harmonic mean size: 0.600 to 0.850mm,% <0.355mm: < 1.0%,%>1.180mm: <5.0%
C3H7BrO
MFCD00145841
Brown-Gray
25g
COCCBr
138.992
Beads
weight total capacity (H):> 4.70 eq/kg
(H): 52.0 to 57.0%
Plastic bottle
(H):> 1.70 eq/L
05,355; 06,302; 09,256
2-bromoethyl methyl ether, 2-methoxyethyl bromide, ethane, 1-bromo-2-methoxy, ether, 2-bromoethyl methyl, methoxyethyl bromide, 2-bromoethylmethylether, bromoethyl methyl ether, 2-bromoethylmethyl ether, 2-bromoethylmethyl-ether, 2-bromo-1-methoxyethane
YZUPZGFPHUVJKC-UHFFFAOYSA-N
1-bromo-2-methoxyethane
80972