missing translation for 'onlineSavingsMsg'
Learn More

Amberlite™ XAD-4, Thermo Scientific Chemicals

Catalog Number 11956991
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy
9003-69-4
C10H10
0.00
MFCD00132704
MYRTYDVEIRVNKP-UHFFFAOYSA-N
divinylbenzene, 1,2-divinylbenzene, o-divinylbenzene, benzene, diethenyl, benzene, 1,2-diethenyl, divinylbenzen, divinyl benzen, divinyl-benzene, poly divinylbenzene, divinylbenzene dvb
66666
1,2-diethenylbenzene
*
This item is not returnable. View return policy
9003-69-4
C10H10
0.00
MFCD00132704
MYRTYDVEIRVNKP-UHFFFAOYSA-N
divinylbenzene, 1,2-divinylbenzene, o-divinylbenzene, benzene, diethenyl, benzene, 1,2-diethenyl, divinylbenzen, divinyl benzen, divinyl-benzene, poly divinylbenzene, divinylbenzene dvb
66666
1,2-diethenylbenzene
*

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Ion exchange resin
TRUSTED_SUSTAINABILITY
Physical Form Crystalline Powder or Crystals
Particle Size 2.0 (uniformity coefficient) Max. 0.490 to 0.690mm (harmonic mean size) 5.0% Max. (<0.300mm) 5.0% Max. (>1.180 mm)
Packaging Plastic bottle
Water Retention Capacity moisture holding capacity: 54.0 to 60.0%
Color White
Quantity 2.5 kg
Chemical Name or Material Amberlite™ XAD-4

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'