missing translation for 'onlineSavingsMsg'
Learn More

Amberlite™ IRN-78 ion-exchange resin, OH-form, ACROS Organics™

Brand:  Acros Organics 429290025

Code : 29

Additional Details : CAS Number : 11128-95-3 Weight : 2.55000kg


Quantity 2.5kg
Packaging Plastic bottle

 View more versions of this product

Product Code. 10545090

  • £205.00 / 2.50kg


EU Stock 1
Add to basket


Specifications

Chemical Identifiers

Amberlite™ IRN-78 ion-exchange resin
Ion-exchange Resin, OH-form
11128-95-3
MFCD00145822

Specifications

Amberlite™ IRN-78 ion-exchange resin
11128-95-3
Solution
Colorless to yellow
Ion-exchange Resin, OH-form
Plastic bottle
MFCD00145822
2.5kg
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Amberlite(TM) IRN78 resin, hydroxide form

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1

Hazard Statement
  • H318-Causes serious eye damage.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS