missing translation for 'onlineSavingsMsg'
Learn More

Amberlite™ IRA-96, ion exchange resin, Thermo Scientific Chemicals

Catalog Number p-7052409
Click to view available options
Quantity:
250 g
1000 g
This item is not returnable. View return policy
39409-19-3
MFCD00166430
This item is not returnable. View return policy
39409-19-3
MFCD00166430

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Physical Form Beads
Color Brown
Quantity 250 g
Chemical Name or Material Amberlite™ IRA-96 ion exchange resin

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'