missing translation for 'onlineSavingsMsg'
Learn More

Amberlite™ IR-120 Na ion-exchange resin, ACROS Organics™

Brand:  Acros Organics 208412500

Code : 29

Additional Details : CAS Number : 9002-23-7 Weight : 0.30000kg

Quantity 250g
Packaging Plastic bottle

 View more versions of this product

Product Code. 10421061

  • £30.60 / 250g


EU Stock 3
Add to basket

Description

Description

Ion exchange resin
Specifications

Chemical Identifiers

Amberlite™ IR120
9002-23-7
MFCD00132707

Specifications

Amberlite™ IR120
(Na+): 45 to 50%
Plastic bottle
(Na+):> 2.0 eq/L
Amber to brown
14, 382
9002-23-7
Particle size analysis: uniformity coefficient: <1.9, harmonic mean size: 0.600 to 0.800mm,% <0.300mm: <2.0%
Beads
MFCD00132707
01,511; 04,266; 05,355; 06,302; 09,256
250g
Videos
SDS