Learn More
Amberlite™ FPC3500(H), ion exchange resin, Thermo Scientific™
Brand: Thermo Scientific Alfa Aesar 045903.A1
Code : NEW
Additional Details : Weight : 1.00000kg
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
- Suitable for the neutralization of strong bases, the recovery of metallic ions, and the isolation and concentration of antibiotics, basic amino acids, enzymes, and peptides
Specifications
Amberlite™ FPC3500 (H) ion exchange resin | |
1.2 g/mL | |
14,832 | |
Insoluble in water. |
MFCD08460528 | |
White | |
1 kg |