missing translation for 'onlineSavingsMsg'
Learn More

Duran™ Amber Wide Neck Glass Reagent Bottles with Flat Head Stopper

Brand:  Duran™ 211883608

Code : A0

Additional Details : Weight : 2.50000kg

 View more versions of this product

Product Code. 11796858

  • £775.00 / Pack of 10


Estimated Shipment
22-06-2018
Add to basket

Specifications

Specifications

250mL
Amber
Standard Ground Joint, Glass Flat-Head Stopper
34/35
Bottle, Reagent
133mm
ISO 4796-2, USP Standard
Suitable for storage of aggressive media, which could attack plastic parts.
Glass
10Pack
70mm
Wide
Videos