Learn More
Emma Safety Footwear Amber Safety Shoes
Supremely comfortable, sculpted ladies safety shoe. Features SmartTecAir™ technology to remove moisture and perspiration instantly.
Brand:  Emma Safety Footwear 942516-37
Additional Details : Weight : 1.19000kg
 View more versions of this product
Product Code. 15939101
Quantity Price
1 £106.11 / 1 pair
10 £93.13 / 1 pair
Estimated Shipment
18-06-2021
Add to basket
Description
Description
  • S1P-Safety shoe with sole plate for dry work
  • GripForce™ Easy Twist
  • Microfiber construction
  • ESD: Electrostatic Discharge
  • Chrome-free lining
  • Hydro-Tec™ Sanitized Silver
Specifications
Specifications
EN ISO 20345:2011, EN 12568:2010
Black
Hydro-Tec™ Sanitized Silver
37
Laces
Amber
Microfiber
Steel