missing translation for 'onlineSavingsMsg'
Learn More

Duran™ Amber Narrow Neck Reagent Bottles with Flat Head Stopper

Guaranteed thermal shock resistance of 160K. DURAN™ Amber Narrow Neck Reagent Bottles with Flat Head Stopper is leak free with regards to liquid and gaseous media.

Brand:  Duran™ 211681705

Code : A0

Additional Details : Weight : 0.70000kg

 View more versions of this product

Product Code. 11726858

  • £419.00 / Pack of 10


Estimated Shipment
20-02-2018
Add to basket

Specifications

Specifications

50mL
Amber
500°C
Bottle, Reagent
Yes
80mm
ISO 4796-2, USP Standard
Standard Ground Joint, Glass Flat-Head Stopper
10Pack
Yes
42mm
Narrow
Videos