missing translation for 'onlineSavingsMsg'
Learn More

AM 630, Tocris Bioscience™

CB2 selective antagonist/inverse agonist. Restrictions apply. See special conditions on product page.

Brand:  Tocris (Bio-Techne) 112050

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13458593

  • £684.77 / 50mg


Estimated Shipment
04-03-2019
Add to basket

Description

Description

CB2 antagonist/inverse agonist (Ki = 31.2 nM) that displays 165-fold selectivity over CB1 receptors. Behaves as a weak partial/inverse agonist at CB1 receptors.
Specifications

Chemical Identifiers

164178-33-0
504.368
iodopravadoline, 6-iodo-2-methyl-1-2-morpholinoethyl-1h-indol-3-yl 4-methoxyphenyl methanone, 6-iodopravadoline, unii-u1lnj6nbka, u1lnj6nbka, 6-iodo-2-methyl-1-2-morpholin-4-ylethyl indol-3-yl-4-methoxyphenyl methanone, 1-2-morpholin-4-yl ethyl-2-methyl-3-4-methoxybenzoyl-6-iodoindole, 1-2-morpholin-4-yl ethyl-2-methyl-3-4-methoxybenzoyl-6-iodoindole, 6-iodo-3-4-methoxybenzoyl-2-methyl-1-2-morpholin-4-yl ethyl indole, tocris-1120
[6-iodo-2-methyl-1-(2-morpholin-4-ylethyl)indol-3-yl]-(4-methoxyphenyl)methanone
C23H25IN2O3
JHOTYHDSLIUKCJ-UHFFFAOYSA-N
4302963
CC1=C(C2=C(N1CCN3CCOCC3)C=C(C=C2)I)C(=O)C4=CC=C(C=C4)OC

Specifications

AM 630
164178-33-0
iodopravadoline, 6-iodo-2-methyl-1-2-morpholinoethyl-1h-indol-3-yl 4-methoxyphenyl methanone, 6-iodopravadoline, unii-u1lnj6nbka, u1lnj6nbka, 6-iodo-2-methyl-1-2-morpholin-4-ylethyl indol-3-yl-4-methoxyphenyl methanone, 1-2-morpholin-4-yl ethyl-2-methyl-3-4-methoxybenzoyl-6-iodoindole, 1-2-morpholin-4-yl ethyl-2-methyl-3-4-methoxybenzoyl-6-iodoindole, 6-iodo-3-4-methoxybenzoyl-2-methyl-1-2-morpholin-4-yl ethyl indole, tocris-1120
CC1=C(C2=C(N1CCN3CCOCC3)C=C(C=C2)I)C(=O)C4=CC=C(C=C4)OC
504.368
504.37
50mg
C23H25IN2O3
JHOTYHDSLIUKCJ-UHFFFAOYSA-N
[6-iodo-2-methyl-1-(2-morpholin-4-ylethyl)indol-3-yl]-(4-methoxyphenyl)methanone
4302963
>98%
Videos