missing translation for 'onlineSavingsMsg'
Learn More

AM 404, Tocris Bioscience™

Anandamide transport inhibitor

Brand:  Tocris (Bio-Techne) 1116/50

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13458573

  • £336.61 / 50mg


Estimated Shipment
04-03-2019
Add to basket

Description

Description

Competitive and selective inhibitor of carrier-mediated anandamide transport (IC50 = 1 μM). Does not activate CB1 receptors or inhibit anandamide hydrolysis but has been shown to activate native and cloned vanilloid receptors (pEC50 = 7.4). Active in vivo. Also available in water soluble emulsion (Cat. No. 1685).
Specifications

Chemical Identifiers

198022-70-7
395.587
unii-xvj94h0u21, chembl39878, n-4-hydroxyphenyl arachidonoyl amide, 5z,8z,11z,14z-n-4-hydroxyphenyl icosa-5,8,11,14-tetraenamide, n-4-hydroxyphenyl-eicosa-5,8,11,14-tetraenamide, n-4-hydroxyphenyl-arachidonylamide, n-4-hydroxyphenyl arachidonylamide, tocris-1116, 5,8,11,14-eicosatetraenamide, n-4-hydroxyphenyl-, all-z, n-4-hydroxyphenyl icosa-5,8,11,14-tetraenamide
(5Z,8Z,11Z,14Z)-N-(4-hydroxyphenyl)icosa-5,8,11,14-tetraenamide
C26H37NO2
IJBZOOZRAXHERC-DOFZRALJSA-N
6604822
CCCCCC=CCC=CCC=CCC=CCCCC(=O)NC1=CC=C(C=C1)O

Specifications

AM 404
198022-70-7
unii-xvj94h0u21, chembl39878, n-4-hydroxyphenyl arachidonoyl amide, 5z,8z,11z,14z-n-4-hydroxyphenyl icosa-5,8,11,14-tetraenamide, n-4-hydroxyphenyl-eicosa-5,8,11,14-tetraenamide, n-4-hydroxyphenyl-arachidonylamide, n-4-hydroxyphenyl arachidonylamide, tocris-1116, 5,8,11,14-eicosatetraenamide, n-4-hydroxyphenyl-, all-z, n-4-hydroxyphenyl icosa-5,8,11,14-tetraenamide
CCCCCC=CCC=CCC=CCC=CCCCC(=O)NC1=CC=C(C=C1)O
395.587
395.58
50mg
C26H37NO2
IJBZOOZRAXHERC-DOFZRALJSA-N
(5Z,8Z,11Z,14Z)-N-(4-hydroxyphenyl)icosa-5,8,11,14-tetraenamide
6604822
>98%
Videos