missing translation for 'onlineSavingsMsg'
Learn More

AM 404 (in Tocrisolve 100), Tocris Bioscience™

Brand:  Tocris (Bio-Techne) 168525

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 25mg

 View more versions of this product

Product Code. 13439913

  • £448.35 / 25mg

Estimated Shipment date
from Supplier 20-12-2019

Please to purchase this item. Need a web account? Register with us today!


Specifications

Chemical Identifiers

198022-70-7
395.587
unii-xvj94h0u21, chembl39878, n-4-hydroxyphenyl arachidonoyl amide, 5z,8z,11z,14z-n-4-hydroxyphenyl icosa-5,8,11,14-tetraenamide, n-4-hydroxyphenyl-eicosa-5,8,11,14-tetraenamide, n-4-hydroxyphenyl-arachidonylamide, n-4-hydroxyphenyl arachidonylamide, tocris-1116, 5,8,11,14-eicosatetraenamide, n-4-hydroxyphenyl-, all-z, n-4-hydroxyphenyl icosa-5,8,11,14-tetraenamide
(5Z,8Z,11Z,14Z)-N-(4-hydroxyphenyl)icosa-5,8,11,14-tetraenamide
C26H37NO2
IJBZOOZRAXHERC-DOFZRALJSA-N
6604822
CCCCCC=CCC=CCC=CCC=CCCCC(=O)NC1=CC=C(C=C1)O

Specifications

25mg
198022-70-7
unii-xvj94h0u21, chembl39878, n-4-hydroxyphenyl arachidonoyl amide, 5z,8z,11z,14z-n-4-hydroxyphenyl icosa-5,8,11,14-tetraenamide, n-4-hydroxyphenyl-eicosa-5,8,11,14-tetraenamide, n-4-hydroxyphenyl-arachidonylamide, n-4-hydroxyphenyl arachidonylamide, tocris-1116, 5,8,11,14-eicosatetraenamide, n-4-hydroxyphenyl-, all-z, n-4-hydroxyphenyl icosa-5,8,11,14-tetraenamide
CCCCCC=CCC=CCC=CCC=CCCCC(=O)NC1=CC=C(C=C1)O
395.587
395.58
AM 404 (in Tocrisolve™ 100)
C26H37NO2
IJBZOOZRAXHERC-DOFZRALJSA-N
(5Z,8Z,11Z,14Z)-N-(4-hydroxyphenyl)icosa-5,8,11,14-tetraenamide
6604822
>98%
Videos
Documents