missing translation for 'onlineSavingsMsg'
Learn More

AM 114, Tocris Bioscience™

20S proteasome inhibitor

Brand:  Tocris (Bio-Techne) 256450

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13431484

  • £515.97 / 50mg

Estimated Shipment date
from Supplier 24-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Inhibitor of the chymotrypsin-like activity of the 20S proteasome (IC50 ∽ 1 μM). Displays anticancer activity; inhibits cell growth in human colon cancer HCT116 p53+/+ cells (IC50 = 1.49 μM).
Specifications

Chemical Identifiers

856849-35-9
377.01
3, 5-Bis-[benzylidene-4-boronic acid]-1-methylpiperidin-4-one
[4-[[5-[(4-boronophenyl)methylidene]-1-methyl-4-oxopiperidin-3-ylidene]methyl]phenyl]boronic acid
C20H21B2NO5
SRPIKXGUPAKTIZ-UHFFFAOYSA-N
57369225
B(C1=CC=C(C=C1)C=C2CN(CC(=CC3=CC=C(C=C3)B(O)O)C2=O)C)(O)O

Specifications

50mg
856849-35-9
3, 5-Bis-[benzylidene-4-boronic acid]-1-methylpiperidin-4-one
B(C1=CC=C(C=C1)C=C2CN(CC(=CC3=CC=C(C=C3)B(O)O)C2=O)C)(O)O
377.01
377.01
AM 114
C20H21B2NO5
SRPIKXGUPAKTIZ-UHFFFAOYSA-N
[4-[[5-[(4-boronophenyl)methylidene]-1-methyl-4-oxopiperidin-3-ylidene]methyl]phenyl]boronic acid
57369225
>97%
Videos
Documents