missing translation for 'onlineSavingsMsg'
Learn More

Aluminum wire, 2.0mm (0.08in) dia, annealed, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog Number 11322398
Click to view available options
Quantity:
1 m
25 m
5 m
This item is not returnable. View return policy
7429-90-5
Al
26.98
MFCD00134029
XAGFODPZIPBFFR-UHFFFAOYSA-N
aluminum, metal, powder, aluminio, metana, adom, alumina fibre, aluminum flake, dust, noral aluminum
5359268
CHEBI:33629
[Al]
This item is not returnable. View return policy
7429-90-5
Al
26.98
MFCD00134029
XAGFODPZIPBFFR-UHFFFAOYSA-N
aluminum, metal, powder, aluminio, metana, adom, alumina fibre, aluminum flake, dust, noral aluminum
5359268
CHEBI:33629
[Al]

Form: approx. 8.4g/m; Temper: soft

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Silver
Appearance Silver-colored wire
Density 2.7g/cm3
Assay Percent Range 99.999% (Metals basis)
Quantity 5 m
IUPAC Name aluminum
Odor Odorless
Form Wire
Chemical Name or Material Aluminum, Annealed

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'