missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aluminum trifluoromethanesulfonate, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
Description
Aluminum trifluoromethanesulfonate is used in pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | ∼300°C |
Color | White |
Linear Formula | Al(CF3SO3)3 |
Quantity | 10 g |
UN Number | UN3261 |
Sensitivity | Hygroscopic |
Solubility Information | Insoluble in water. |
Formula Weight | 474.2 |
Physical Form | Powder |
Chemical Name or Material | Aluminum trifluoromethanesulfonate |
Safety and Handling
Product Identifier
- Aluminum trifluoromethanesulfonate
- Danger
- Skin corrosion/irritation Category 1 C
- H314-Causes severe skin burns and eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'