missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aluminum titanium oxide, 99.5% (metals basis), Thermo Scientific Chemicals
56.26 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
25 g
100 g
Description
Aluminum titanium oxide is used in refractory industry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Physical Form | Powder |
Quantity | 25 g |
Solubility Information | Insoluble in water. |
Formula Weight | 181.86 |
Percent Purity | 99.5% |
Odor | Odorless |
Assay | (metals basis) |
Chemical Name or Material | Aluminum titanium oxide |
Safety and Handling
Product Identifier
- Aluminum titanium oxide
- Warning
- Specific target organ toxicity Category 3
- H335-May cause respiratory irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P405-Store locked up.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
- P501-Dispose of contents/container to .
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'