missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aluminum tert-butoxide, 97%, Thermo Scientific Chemicals
26.73 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
5 g
25 g
100 g
Description
Aluminum tert-butoxide is utilized as a reagent in the Oppenauer oxidation of alcohols to ketones. It is also used in Meerwein-Ponndorf-Verley reduction of ketones. It is actively involved in the preparation of 1,1,2-triethoxyethene by reacting with 1,1,2,2-tetraethoxyethane followed by the elimination of alcohol.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 200°C to 202°C |
Density | 1.0251 g/mL |
Boiling Point | 156°C (2 mmHg) |
Odor | Alcohol-like |
Quantity | 5 g |
UN Number | UN2925 |
Beilstein | 4148229 |
Sensitivity | Moisture sensitive |
Merck Index | 14,333 |
Solubility Information | Very soluble in organic solvents like alcohols,benzene,toluene and xylene. |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'