missing translation for 'onlineSavingsMsg'
Learn More

Aluminum sulfate hydrate, Puriss., meets analytical spec. of BP, Ph. Eur., 100-110%, 51.0-59.0% Al2(SO4)3 basis, Honeywell Fluka™
BIOPHARMA

Catalog Number p-7071760
Click to view available options
Quantity:
1 kg
2.5 kg
20 kg
25 kg
6 x 1 kg
7784-31-8
Al2H36O30S3
666.401
AMVQGJHFDJVOOB-UHFFFAOYSA-H
aluminum sulfate octadecahydrate, patent alum, cake alum octadecahydrate, unii-tcs9l00g8f, dialuminum sulfate octadecahydrate, tcs9l00g8f, aluminum sulfate.18h2o, aluminum sulfate.18h2o, dialuminium sulfate octadecahydrate, aluminum sulfate-water 1/18
22377415
CHEBI:74779
dialuminum;trisulfate;octadecahydrate
O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Al+3].[Al+3]
7784-31-8
Al2H36O30S3
666.401
AMVQGJHFDJVOOB-UHFFFAOYSA-H
aluminum sulfate octadecahydrate, patent alum, cake alum octadecahydrate, unii-tcs9l00g8f, dialuminum sulfate octadecahydrate, tcs9l00g8f, aluminum sulfate.18h2o, aluminum sulfate.18h2o, dialuminium sulfate octadecahydrate, aluminum sulfate-water 1/18
22377415
CHEBI:74779
dialuminum;trisulfate;octadecahydrate
O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Al+3].[Al+3]

Puriss., meets analytical specification of BP, Ph. Eur., 100-110%, 51.0-59.0% Al2(SO4)3 basis

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Quantity 1 kg
Linear Formula Al2(SO4)3 · 18H2O
UN Number 3260
Formula Weight 342.15g/mol (anhydrous basis)
Percent Purity 100 to 110%
Grade puriss.
Chemical Name or Material Aluminum Sulfate Hydrate, Meets analytical specification of Ph.Eur.BP
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'