missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aluminum sulfate hydrate, ACS, 98.0-102%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 g
1 kg
Description
Aluminum sulfate is a chemical compound commonly used as a flocculating agent in the purification of drinking water and waste water treatment plants, and also in paper manufacturing. Used as coagulant in decolorization and COD removal of secondary yeast wastewater effluents. It is also plays a significant role in arsenic removal by coagulation process.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
pH | 3.5 |
Physical Form | Crystalline |
Quantity | 250 g |
Merck Index | 14,366 |
Solubility Information | It is slightly soluble in alcohol and dilute mineral acids. |
Formula Weight | 342.15 (Anhydrous) |
Percent Purity | 98.0 to 102% |
Grade | ACS |
Odor | Odorless |
Density | 1.61 g/mL |
Show More |
Safety and Handling
Product Identifier
- Aluminum sulfate hydrate
- Danger
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 1
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H335-May cause respiratory irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P321-Specific treatment (see on this label).
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P362+P364-Take off contaminated clothing and wash it before reuse.
- P405-Store locked up.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
- P501-Dispose of contents/container to .
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'