missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aluminum Solution for ICP, 1g/L HCl matrix, Fisher Chemical™
Click to view available options
Quantity:
1 mL
100 mL
250 mL
Specifications
Concentration | 100μg/mL |
Packaging | Ampoule |
Quantity | 1 mL |
Type | Pesticide and Metabolite Standards |
Analyte or Component Names | Alloxydine |
Solvent or Matrix | Acetonitrile |
Chemical Name or Material | Aminohippuric acid reagent. |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'