missing translation for 'onlineSavingsMsg'
Learn More

Aluminum shot, ACS, 8-12mm (0.31-0.47in), Thermo Scientific Chemicals

Catalog Number p-3256864
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy
7429-90-5
Al
26.98
MFCD00134029
XAGFODPZIPBFFR-UHFFFAOYSA-N
aluminum, metal, powder, aluminio, metana, adom, alumina fibre, aluminum flake, dust, noral aluminum
5359268
CHEBI:33629
aluminium
[Al]
This item is not returnable. View return policy
7429-90-5
Al
26.98
MFCD00134029
XAGFODPZIPBFFR-UHFFFAOYSA-N
aluminum, metal, powder, aluminio, metana, adom, alumina fibre, aluminum flake, dust, noral aluminum
5359268
CHEBI:33629
aluminium
[Al]

Typically 3-4mm thick

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 2.7
Quantity 100 g
Formula Weight 26.98
Grade ACS
Form Flattened Shot
Chemical Name or Material Aluminum

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'