missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey Nagel™ Standard SIL G Silica Layers on Alugram™ Aluminum Sheets UV254
Separate non-volatile mixtures with unmodified standard silica layers for TLC (Thin-Layer Chromatography). Standard SIL G Silica Layers on Alugram™ Aluminum Sheets UV254 come in a variety of sizes.
Brand: Macherey Nagel™ 818160
Code : NEW
Additional Details : Weight : 0.16020kg
Description
Alugram™ Aluminum Sheets for TLC- Pre-coated ready to use plates
- Broad range of plate sizes, surface coatings and backings
- Silica 60
- specific surface (BET) ~ 500 m2/g
- Mean pore size 60 Å
- Specific pore volume 0.75 mL/g,
- Particle size 5–17 µm
- Standard grade
- For analytical plates 0.25 mm
- For preparative plates 0.5 and 1 mm
- For 2 mm preparative layers a slightly coarser material is used.
- Manganese activated zinc silicate with green fluorescence for short-wave UV (254 nm)
- Special inorganic fluorescent pigment with blue fluorescence for long-wave UV (366 nm)
- Highly polymeric products, which are stable in almost all organic solvents and resistant towards aggressive visualisation reagents