missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Aluminum sec-butoxide, 95%

Brand:  Alfa Aesar™ 030721.22

Code : NEW

Additional Details : CAS Number : 2269-22-9 Weight : 0.10000kg Transport : UN number : 1992 Chem class : 3 Pack group : III

Quantity 100g

 View more versions of this product

Product Code. 11327338

  • £20.10 / 100g


EU Stock 2
Add to basket

Specifications

Chemical Identifiers

Aluminum sec-butoxide
246.32
UN1992
Aluminum tri-sec-butoxide
2269-22-9
MFCD00009327
3910908

Specifications

Aluminum sec-butoxide
2269-22-9
27° (82°F)
Alcohol-like
Moisture Sensitive
95%
MFCD00009327
3910908
Hydrolyzes in water; Soluble in aromatic hydrocarbons, alcohols
200° to 206°C (30mmHg)
100g
0.967g/mL
1.4390
Liquid
246.32
UN1992
Aluminum tri-sec-butoxide
Videos
SDS