missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Alfa Aesar Aluminum rod, alloy 6061, 19.0mm (0.75 in.) dia.

Brand:  Thermo Scientific Alfa Aesar 042054.BV

Aluminum rod, alloy 6061, 19.0mm (0.75 in.) dia.

Code : NEW

Additional Details : Weight : 0.01000kg

Product Code. 11329717

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.