missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Aluminum rod, alloy 6061, 19.0mm (0.75 in.) dia.

Brand:  Alfa Aesar™ 042054.CE

Code : NEW

Additional Details : Weight : 0.01000kg

Quantity 5 x 30cm

 View more versions of this product

Product Code. 11319717

  • £68.79 / Pack of 5


Estimated Shipment date
from Supplier 12-06-2018
Add to basket

Specifications

Chemical Identifiers

Aluminum rod, Alloy 6061
19.0mm (0.75 in.) dia.
MFCD00214039

Specifications

Aluminum rod, Alloy 6061
5 x 30cm
Rod
Odorless
≈7.69g/cm
19.0mm (0.75 in.) dia.
MFCD00214039
Videos
SDS