missing translation for 'onlineSavingsMsg'
Learn More

Bochem™ Aluminum Retort Clamp

Bochem™ Aluminum Retort Clamp features three, PVC coated prongs.

Brand:  Bochem™ 5566

Code : A0

Additional Details : Weight : 0.80000kg

Product Code. 10771592

  • £154.00 / Each
Estimated Shipment: 13-06-2024
to see stock.
Add to basket

Description

Description

  • Aluminum
  • 3-prong
  • PVC coated fingers
Specifications

Specifications

Aluminum
0 to 125 mm
10 mm
PVC Coated Prongs
130 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers